Product Details

SNP ID
rs4819631
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:17777838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTCTCTCCCTTTCCACCTTCTG[C/T]TCTGAGCCACCCGTCCCCTCCAGAA
Phenotype
MIM: 601997 MIM: 608882
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BID PubMed Links

Gene Details

Gene
BID
Gene Name
BH3 interacting domain death agonist
There are no transcripts associated with this gene.

Gene
LINC00528
Gene Name
long intergenic non-protein coding RNA 528
There are no transcripts associated with this gene.

Gene
MICAL3
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 3
There are no transcripts associated with this gene.

View Full Product Details