Product Details
- SNP ID
-
rs4940456
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:59296510 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATAACTAAAAGGAAAGTAAGGTTCC[C/T]GCTGCAAGGTGTTAGCTAAATGCTC
- Phenotype
-
MIM: 609586
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CPLX4
PubMed Links
Gene Details
- Gene
- CPLX4
- Gene Name
- complexin 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_181654.3 |
858 |
UTR 3 |
|
|
NP_857637.1 |
View Full Product Details