Product Details

SNP ID
rs4995113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4797657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAAGGTTGGGGCGCTCTGAGGG[C/T]GTGGCAAGGACCCTTAGAAGTCCTT
Phenotype
MIM: 610660 MIM: 614574 MIM: 611562
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLYR1 PubMed Links

Gene Details

Gene
GLYR1
Gene Name
glyoxylate reductase 1 homolog
There are no transcripts associated with this gene.

Gene
ROGDI
Gene Name
rogdi homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024589.2 Intron NP_078865.1
XM_006720947.3 Intron XP_006721010.1
XM_006720948.3 Intron XP_006721011.1
Gene
SEPT12
Gene Name
septin 12
There are no transcripts associated with this gene.

Gene
SMIM22
Gene Name
small integral membrane protein 22
There are no transcripts associated with this gene.

View Full Product Details