Product Details

SNP ID
rs4326571
Assay Type
Functionally tested
NCBI dbSNP Submissions
47
Location
Chr.1:1071610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGGGGATCCTCTTGTCCACCCC[G/A]TCAGGACCCAGCCTGGAGAATGAGG
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
LOC100288175 PubMed Links

Gene Details

Gene
LOC100288175
Gene Name
uncharacterized LOC100288175
There are no transcripts associated with this gene.

Gene
LOC105378948
Gene Name
uncharacterized LOC105378948
There are no transcripts associated with this gene.

Gene
RNF223
Gene Name
ring finger protein 223
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205252.1 1258 Intron NP_001192181.1
XM_017001286.1 1258 UTR 3 XP_016856775.1
XM_017001287.1 1258 Intron XP_016856776.1

View Full Product Details