Product Details

SNP ID
rs4481671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:93065826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCATTCATTCACTCATTCCTGCA[A/C]TGATGTCTCCAGATATGGAGCTTGG
Phenotype
MIM: 616429
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SUSD3 PubMed Links

Gene Details

Gene
SUSD3
Gene Name
sushi domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287005.1 Intron NP_001273934.1
NM_001287006.1 Intron NP_001273935.1
NM_001287007.1 Intron NP_001273936.1
NM_001287008.1 Intron NP_001273937.1
NM_145006.3 Intron NP_659443.1

View Full Product Details