Product Details

SNP ID
rs4767898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120456707 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTATTCTGAGGATGTGCTACTCAC[C/G]CAGGCTGGACTAGGTACCCTTTCTA
Phenotype
MIM: 601943
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GATC PubMed Links

Gene Details

Gene
GATC
Gene Name
glutamyl-tRNA amidotransferase subunit C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176818.2 Intron NP_789788.1
Gene
SRSF9
Gene Name
serine and arginine rich splicing factor 9
There are no transcripts associated with this gene.

View Full Product Details