Product Details

SNP ID
rs4981800
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:30898572 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTCAGATGGACCATGAATAGAGA[A/C]CACAATGCTGGACCAAGTTGTATGT
Phenotype
MIM: 603196 MIM: 614766
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
COCH PubMed Links

Gene Details

Gene
COCH
Gene Name
cochlin
There are no transcripts associated with this gene.

Gene
LOC100506071
Gene Name
uncharacterized LOC100506071
There are no transcripts associated with this gene.

Gene
STRN3
Gene Name
striatin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083893.1 Intron NP_001077362.1
NM_014574.3 Intron NP_055389.3
XM_005267569.3 Intron XP_005267626.1
XM_005267570.3 Intron XP_005267627.1

View Full Product Details