Product Details

SNP ID
rs4790649
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4584166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCCGCGGAGTGGGGCGGCAGCT[C/G]TGGGAGCTGTAGGTGTCCCCATTAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SMTNL2 PubMed Links

Gene Details

Gene
SMTNL2
Gene Name
smoothelin like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114974.1 Intron NP_001108446.1
NM_198501.2 Intron NP_940903.2
XM_005256622.3 Intron XP_005256679.1
XM_006721515.2 Intron XP_006721578.1

View Full Product Details