Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001324282.1 | Intron | NP_001311211.1 | |||
NM_001324283.1 | Intron | NP_001311212.1 | |||
NM_001324284.1 | Intron | NP_001311213.1 | |||
NM_001324285.1 | Intron | NP_001311214.1 | |||
NM_001324286.1 | Intron | NP_001311215.1 | |||
NM_001324287.1 | Intron | NP_001311216.1 | |||
NM_001324288.1 | Intron | NP_001311217.1 | |||
NM_001324289.1 | Intron | NP_001311218.1 | |||
NM_003473.3 | Intron | NP_003464.1 | |||
XM_011519695.2 | Intron | XP_011517997.1 | |||
XM_017016685.1 | Intron | XP_016872174.1 |