Product Details

SNP ID
rs4695238
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:47486414 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAACAAGTCTGTATGGACAGAGC[A/G]AACCTCTAACTCAAAATCAACCTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP10D PubMed Links

Gene Details

Gene
ATP10D
Gene Name
ATPase phospholipid transporting 10D (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020453.3 Intron NP_065186.3
XM_005248119.4 Intron XP_005248176.1
XM_005248120.4 Intron XP_005248177.1
XM_011513722.2 Intron XP_011512024.1
XM_017008472.1 Intron XP_016863961.1

View Full Product Details