Product Details

SNP ID
rs4233426
Assay Type
Functionally tested
NCBI dbSNP Submissions
44
Location
Chr.1:89410097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCGTCCTCCTGGATCCCACAACA[C/T]TGATAAGTCACTGTCATGCTAACAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GBP1P1 PubMed Links

Gene Details

Gene
GBP1P1
Gene Name
guanylate binding protein 1 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details