Product Details

SNP ID
rs4519167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:7089601 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGGCTCACCCTGGCAGGAATGC[C/G]TGTCTTCCTGAAGCTCATAGCCTGG
Phenotype
MIM: 613785 MIM: 608974
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C1R PubMed Links

Gene Details

Gene
C1R
Gene Name
complement C1r subcomponent
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001733.4 690 Missense Mutation ACG,AGG T186R NP_001724.3
XM_017019915.1 690 Missense Mutation ACG,AGG T200R XP_016875404.1
Gene
C1RL
Gene Name
complement C1r subcomponent like
There are no transcripts associated with this gene.

View Full Product Details