Product Details

SNP ID
rs4971074
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:155321741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTGAGAGACCCTACCCTTCTGG[G/T]TCTCTAGAAGCCATCCCATCGCCGC
Phenotype
MIM: 134629
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FDPS PubMed Links

Gene Details

Gene
FDPS
Gene Name
farnesyl diphosphate synthase
There are no transcripts associated with this gene.

Gene
LOC105371451
Gene Name
uncharacterized LOC105371451
There are no transcripts associated with this gene.

Gene
RUSC1
Gene Name
RUN and SH3 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105203.1 197 UTR 5 NP_001098673.1
NM_001105204.1 197 UTR 5 NP_001098674.1
NM_001105205.1 197 Intron NP_001098675.1
NM_001278227.1 197 Intron NP_001265156.1
NM_001278228.1 197 Intron NP_001265157.1
NM_001278229.1 197 Intron NP_001265158.1
NM_001278230.1 197 Intron NP_001265159.1
NM_014328.4 197 Intron NP_055143.2
XM_006711254.1 197 UTR 5 XP_006711317.1
XM_006711256.1 197 UTR 5 XP_006711319.1
XM_006711257.1 197 Intron XP_006711320.1
XM_017000891.1 197 UTR 5 XP_016856380.1
XM_017000892.1 197 Intron XP_016856381.1
Gene
RUSC1-AS1
Gene Name
RUSC1 antisense RNA 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039517.1 197 Intron NP_001034606.1

View Full Product Details