Product Details

SNP ID
rs4970524
Assay Type
Functionally tested
NCBI dbSNP Submissions
42
Location
Chr.1:32477061 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCTCATTTACCTCTATGCCTAAG[C/T]ACGTGTTCTTCCTCTGCCCAAGATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZBTB8B PubMed Links

Gene Details

Gene
ZBTB8B
Gene Name
zinc finger and BTB domain containing 8B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145720.1 Intron NP_001139192.1

View Full Product Details