Product Details
- SNP ID
-
hCV28001087
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:29721690 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGCCCAGGGTGAGGGCAGATCTT[A/C]GAGGAGTCAGAATGGGTCGGGGGAA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CABP7
PubMed Links
Gene Details
- Gene
- CABP7
- Gene Name
- calcium binding protein 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_182527.2 |
|
Intron |
|
|
NP_872333.1 |
- Gene
- ZMAT5
- Gene Name
- zinc finger matrin-type 5
There are no transcripts associated with this gene.
View Full Product Details