Product Details

SNP ID
rs4720089
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:32964026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCAGTCAGGGGGGCCTGACAGGG[G/T]CAGTGGTGTCAGAACAACTGGCCAG
Phenotype
MIM: 616257
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FKBP9 PubMed Links

Gene Details

Gene
FKBP9
Gene Name
FK506 binding protein 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284341.1 Intron NP_001271270.1
NM_001284343.1 Intron NP_001271272.1
NM_007270.4 Intron NP_009201.2
XM_011515115.2 Intron XP_011513417.1

View Full Product Details