Product Details

SNP ID
rs4944839
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73403697 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTACCAGAGGCACATGACTGTCCC[C/T]GCTCTCCTGGGCTCAGTCCTTTTGC
Phenotype
MIM: 616316 MIM: 611211
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM168A PubMed Links

Gene Details

Gene
FAM168A
Gene Name
family with sequence similarity 168 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286050.1 3666 UTR 3 NP_001272979.1
NM_001286051.1 3666 UTR 3 NP_001272980.1
NM_015159.2 3666 UTR 3 NP_055974.1
XM_017017415.1 3666 UTR 3 XP_016872904.1
XM_017017416.1 3666 UTR 3 XP_016872905.1
Gene
RELT
Gene Name
RELT tumor necrosis factor receptor
There are no transcripts associated with this gene.

View Full Product Details