Product Details

SNP ID
hCV28030044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:1354406 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACAGTGACCAGTAGCAGGATGAA[T/G]AGCAGCAGCGTGGCCAGCACGTAGC
Phenotype
MIM: 601365
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
DVL1 PubMed Links

Gene Details

Gene
DVL1
Gene Name
dishevelled segment polarity protein 1
There are no transcripts associated with this gene.

Gene
MXRA8
Gene Name
matrix remodeling associated 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282582.1 1343 Silent Mutation CTA,CTC L351L NP_001269511.1
NM_001282583.1 1343 Silent Mutation CTA,CTC L342L NP_001269512.1
NM_001282584.1 1343 Silent Mutation CTA,CTC L250L NP_001269513.1
NM_001282585.1 1343 Silent Mutation CTA,CTC L351L NP_001269514.1
NM_032348.3 1343 Silent Mutation CTA,CTC L351L NP_115724.1

View Full Product Details