Product Details

SNP ID
rs4878379
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:27245797 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTACAGAATATTTTTTAAAGTGTTA[G/T]ACATAAGACAACCATAATCTCTGAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00032 PubMed Links

Gene Details

Gene
LINC00032
Gene Name
long intergenic non-protein coding RNA 32
There are no transcripts associated with this gene.

View Full Product Details