Product Details

SNP ID
rs8069166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:443347 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCGGGTGGGCCTGTGGCGTGGCTC[A/G]AAGCGCACCTCGGCCTTGTAGTTGC
Phenotype
MIM: 615928
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM101B PubMed Links

Gene Details

Gene
FAM101B
Gene Name
family with sequence similarity 101 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182705.2 526 Silent Mutation TTC,TTT F154F NP_874364.1

View Full Product Details