Product Details
- SNP ID
-
rs7298338
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:55246805 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCAAGAGATGTTTAAATCAACTTT[C/G]CTTTGATCTTAAAAGACAGAGTTCT
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR6C74
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117172475] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR6C74
- Gene Name
- olfactory receptor family 6 subfamily C member 74
View Full Product Details