Product Details
- SNP ID
-
rs7179643
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:84817811 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCATGCGCCTCAGGCCTCTGGCAAT[G/T]CGAGGCTCAAGAGAGGACAAAGGCT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ALPK3
PubMed Links
Gene Details
- Gene
- ALPK3
- Gene Name
- alpha kinase 3
View Full Product Details