Product Details
- SNP ID
-
rs7589313
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:173907183 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAATTTGAGGTATACAGATAATCG[A/C]GTCCACTAAATGTTACAACAGGAAT
- Phenotype
-
MIM: 601804
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SP3
PubMed Links
Gene Details
- Gene
- SP3
- Gene Name
- Sp3 transcription factor
View Full Product Details