Product Details

SNP ID
rs7924894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67010362 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTCTGGAGCGAGCCACCTTGCCC[A/C]GGTCCAGGCTCCAGCTCTAACATGC
Phenotype
MIM: 606436
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SYT12 PubMed Links

Gene Details

Gene
SYT12
Gene Name
synaptotagmin 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177880.1 Intron NP_001171351.1
NM_001318773.1 Intron NP_001305702.1
NM_001318775.1 Intron NP_001305704.1
NM_177963.3 Intron NP_808878.1
XM_006718737.3 Intron XP_006718800.1
XM_011545346.2 Intron XP_011543648.1
XM_011545347.1 Intron XP_011543649.1
XM_017018547.1 Intron XP_016874036.1
XM_017018548.1 Intron XP_016874037.1

View Full Product Details