Product Details

SNP ID
rs7015362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:105322780 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCATGTTGAAAGTAAAATGTTCTG[G/T]TTAGAGAATAATCTTGTTAGCAGTT
Phenotype
MIM: 603693
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZFPM2 PubMed Links

Gene Details

Gene
ZFPM2
Gene Name
zinc finger protein, FOG family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012082.3 Intron NP_036214.2
XM_011516947.2 Intron XP_011515249.1
XM_011516948.2 Intron XP_011515250.1

View Full Product Details