Product Details

SNP ID
rs7829369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:73992134 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTGAAGTTTTTTAGCATAGGGTC[A/T]CATAGAGACAATAATACTCTTAATT
Phenotype
MIM: 605243 MIM: 612418
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LY96 PubMed Links

Gene Details

Gene
LY96
Gene Name
lymphocyte antigen 96
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195797.1 Intron NP_001182726.1
NM_015364.4 Intron NP_056179.3
XM_011517508.2 Intron XP_011515810.1
XM_017013299.1 Intron XP_016868788.1
XM_017013300.1 Intron XP_016868789.1
Gene
TMEM70
Gene Name
transmembrane protein 70
There are no transcripts associated with this gene.

View Full Product Details