Product Details
- SNP ID
-
rs7734383
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:74642029 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCGCCTCCCAGCTTGCTTTAAAAC[A/T]CTAGGCGCAGGAGCGATTTAAACTG
- Phenotype
-
MIM: 605173
MIM: 606873
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ENC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150885044] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ENC1
- Gene Name
- ectodermal-neural cortex 1
- Gene
- HEXB
- Gene Name
- hexosaminidase subunit beta
View Full Product Details