Product Details

SNP ID
rs6894877
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:74643178 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGAGCCAATCTATGACTAGTTTT[C/T]GGGCAGTTGGGCAGTTTTTAATGCT
Phenotype
MIM: 605173 MIM: 606873
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENC1 PubMed Links

Gene Details

Gene
ENC1
Gene Name
ectodermal-neural cortex 1
There are no transcripts associated with this gene.

Gene
HEXB
Gene Name
hexosaminidase subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000521.3 Intron NP_000512.1
NM_001292004.1 Intron NP_001278933.1

View Full Product Details