Product Details

SNP ID
rs6767102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:160267353 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGATCAATCAGAACATTCCATTTT[A/T]TTCTACACCAAGAAGGTATGCTCCC
Phenotype
MIM: 611177
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IFT80 PubMed Links

Gene Details

Gene
IFT80
Gene Name
intraflagellar transport 80
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190241.1 Intron NP_001177170.1
NM_001190242.1 Intron NP_001177171.1
NM_020800.2 Intron NP_065851.1

View Full Product Details