Product Details
- SNP ID
-
rs7844014
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:86524410 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAACGTTAGGGTGACTGGGAGAAT[G/T]ATTGATATATTTGGTGTTTCTAGGG
- Phenotype
-
MIM: 604207
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CPNE3
PubMed Links
Gene Details
- Gene
- CPNE3
- Gene Name
- copine 3
View Full Product Details