Product Details

SNP ID
rs7062457
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:21954690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGGGAAGGCAGAGGGACCTCCT[C/T]AGCGACATGTGTATCAAGTACGGTC
Phenotype
MIM: 300105
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMS PubMed Links

Gene Details

Gene
SMS
Gene Name
spermine synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258423.1 Intron NP_001245352.1
NM_004595.4 Intron NP_004586.2
XM_005274582.2 Intron XP_005274639.1
XM_011545568.2 Intron XP_011543870.1
XM_017029753.1 Intron XP_016885242.1
XM_017029754.1 Intron XP_016885243.1
XM_017029755.1 Intron XP_016885244.1

View Full Product Details