Product Details

SNP ID
rs7686994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:145492399 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTGCTAGAATGGCTTACAGAGTT[C/T]GGGGAAACTTACATTTACTGATTTA
Phenotype
MIM: 601595
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMAD1 PubMed Links

Gene Details

Gene
SMAD1
Gene Name
SMAD family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003688.1 Intron NP_001003688.1
NM_005900.2 Intron NP_005891.1
XM_005262992.3 Intron XP_005263049.1
XM_006714217.2 Intron XP_006714280.1
XM_011531961.1 Intron XP_011530263.1
XM_011531962.1 Intron XP_011530264.1
XM_011531963.1 Intron XP_011530265.1
XM_011531964.1 Intron XP_011530266.1
Gene
SMAD1-AS2
Gene Name
SMAD1 antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details