Product Details

SNP ID
rs4235707
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:78373769 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTTTACCTCTATGGTATCTGATA[A/G]CATTTGGTATCTAATTCTTTTGCCA
Phenotype
MIM: 606911
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCAMP1 PubMed Links

Gene Details

Gene
SCAMP1
Gene Name
secretory carrier membrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290229.1 Intron NP_001277158.1
NM_004866.5 Intron NP_004857.4
XM_011543727.2 Intron XP_011542029.1

View Full Product Details