Product Details

SNP ID
rs6662272
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:45507452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTACTCAGCACGCCTGCCATGTTT[C/G]ACCGGGCCCTCAAGCCCTTCTTGCA
Phenotype
MIM: 609831 MIM: 176763
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC163P PubMed Links

Gene Details

Gene
CCDC163P
Gene Name
coiled-coil domain containing 163, pseudogene
There are no transcripts associated with this gene.

Gene
MMACHC
Gene Name
methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015506.2 236 Missense Mutation CAC,GAC H60D NP_056321.2
XM_005270724.4 236 Intron XP_005270781.1
XM_011541204.2 236 Missense Mutation CAC,GAC H3D XP_011539506.1
Gene
PRDX1
Gene Name
peroxiredoxin 1
There are no transcripts associated with this gene.

View Full Product Details