Product Details

SNP ID
rs6656471
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:201405719 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACAGCTGAGCTTGGCCTCCTTGC[A/G]CCGTGGGGACCGGGCACCCTGTGGG
Phenotype
MIM: 602314 MIM: 191042
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LAD1 PubMed Links

Gene Details

Gene
LAD1
Gene Name
ladinin 1
There are no transcripts associated with this gene.

Gene
TNNI1
Gene Name
troponin I1, slow skeletal type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003281.3 4191 UTR 3 NP_003272.3

View Full Product Details