Product Details

SNP ID
rs6971372
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:54547578 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTAAACTTATAGTTCCACTCTCT[C/T]GCTTATTACTTTTAATAAGTGTGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VSTM2A PubMed Links

Gene Details

Gene
VSTM2A
Gene Name
V-set and transmembrane domain containing 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301009.1 Intron NP_001287938.1
NM_001317843.1 Intron NP_001304772.1
NM_182546.3 Intron NP_872352.2
XM_006715665.3 Intron XP_006715728.1
XM_006715666.3 Intron XP_006715729.1
XM_017011841.1 Intron XP_016867330.1
Gene
VSTM2A-OT1
Gene Name
VSTM2A overlapping transcript 1
There are no transcripts associated with this gene.

View Full Product Details