Product Details

SNP ID
rs7723592
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138344541 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAATGTTTTTGATTCCAGGTAGTT[G/T]GTCACAATGGAGGCAATAGAGCATA
Phenotype
MIM: 157680 MIM: 609372 MIM: 609373
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CDC25C PubMed Links

Gene Details

Gene
CDC25C
Gene Name
cell division cycle 25C
There are no transcripts associated with this gene.

Gene
FAM53C
Gene Name
family with sequence similarity 53 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135647.1 Intron NP_001129119.1
NM_016605.2 Intron NP_057689.1
Gene
KDM3B
Gene Name
lysine demethylase 3B
There are no transcripts associated with this gene.

View Full Product Details