Product Details

SNP ID
rs6993871
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:29334187 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCATCCTGTCTTTCATGGCTAATA[C/G]CAAAGTTGACAATCAATGAGGGTCA
Phenotype
MIM: 602747
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DUSP4 PubMed Links

Gene Details

Gene
DUSP4
Gene Name
dual specificity phosphatase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001394.6 7056 UTR 3 NP_001385.1
NM_057158.3 7056 UTR 3 NP_476499.1
XM_011544428.2 7056 UTR 3 XP_011542730.1

View Full Product Details