Product Details

SNP ID
rs7929700
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5507331 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGAAATTGGGTGTAATTGCATCT[C/G]AAGATAGGCATAAGCAGAGCTTAGA
Phenotype
MIM: 605473
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR51B5 PubMed Links

Gene Details

Gene
OR51B5
Gene Name
olfactory receptor family 51 subfamily B member 5
There are no transcripts associated with this gene.

Gene
UBQLN3
Gene Name
ubiquilin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017481.2 2415 UTR 3 NP_059509.1
XM_011520144.2 2415 UTR 3 XP_011518446.1
XM_011520145.2 2415 UTR 3 XP_011518447.1
Gene
UBQLNL
Gene Name
ubiquilin-like
There are no transcripts associated with this gene.

View Full Product Details