Product Details

SNP ID
rs7710005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:233457 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATCTAGCAATTGTTAGGTAATAA[A/G]TATGTGTGGTTTTTTGCAGGCATAT
Phenotype
MIM: 600857
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SDHA PubMed Links

Gene Details

Gene
SDHA
Gene Name
succinate dehydrogenase complex flavoprotein subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001294332.1 Intron NP_001281261.1
NM_004168.3 Intron NP_004159.2
XM_005248331.3 Intron XP_005248388.1
XM_011514072.1 Intron XP_011512374.1
XM_011514073.1 Intron XP_011512375.1
XM_017009685.1 Intron XP_016865174.1

View Full Product Details