Product Details
- SNP ID
-
rs6482316
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:23702145 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGCAAAAATGGGGCCAACAACCTT[A/G]AAGTGCAGCATATCTATGAGAAATT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA1217
PubMed Links
Gene Details
- Gene
- KIAA1217
- Gene Name
- KIAA1217
View Full Product Details