Product Details

SNP ID
rs6561676
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:52664699 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAATGGCCTGGAAAAGTTAAAAAC[A/C]ATCTATTCATGGTAGTTGTGGGGAG
Phenotype
MIM: 604098
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SUGT1 PubMed Links

Gene Details

Gene
SUGT1
Gene Name
SGT1 homolog, MIS12 kinetochore complex assembly cochaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130912.2 Intron NP_001124384.1
NM_001320831.1 Intron NP_001307760.1
NM_006704.4 Intron NP_006695.1
XM_011534891.2 Intron XP_011533193.1
XM_011534893.2 Intron XP_011533195.1
XM_017020365.1 Intron XP_016875854.1
XM_017020366.1 Intron XP_016875855.1
XM_017020367.1 Intron XP_016875856.1
XM_017020368.1 Intron XP_016875857.1

View Full Product Details