Product Details
- SNP ID
-
rs7390781
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:27111219 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAAACAGCACTGCCTTTAAAAGG[C/G]AGAGGACCAAGGAGAGGAAGTAGGT
- Phenotype
-
MIM: 600221
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TEK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78927199] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TEK
- Gene Name
- TEK receptor tyrosine kinase
View Full Product Details