Product Details

SNP ID
rs7043508
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:27248729 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAATGCAATGTTTTGAAATTTTCT[A/G]GTCTGGAGGACATGCAGGATAGAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00032 PubMed Links

Gene Details

Gene
LINC00032
Gene Name
long intergenic non-protein coding RNA 32
There are no transcripts associated with this gene.

View Full Product Details