Product Details

SNP ID
rs7776981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:45106071 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGGGCCAGTGCCTACCATAGACA[A/G]GGCAGGCACTGAATTTGACATGGCT
Phenotype
MIM: 611335 MIM: 611325
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNORA5A PubMed Links

Gene Details

Gene
SNORA5A
Gene Name
small nucleolar RNA, H/ACA box 5A
There are no transcripts associated with this gene.

Gene
SNORA5B
Gene Name
small nucleolar RNA, H/ACA box 5B
There are no transcripts associated with this gene.

Gene
SNORA5C
Gene Name
small nucleolar RNA, H/ACA box 5C
There are no transcripts associated with this gene.

Gene
TBRG4
Gene Name
transforming growth factor beta regulator 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261834.1 Intron NP_001248763.1
NM_004749.3 Intron NP_004740.2
NM_030900.3 Intron NP_112162.1
NM_199122.2 Intron NP_954573.1

View Full Product Details