Product Details

SNP ID
rs6580046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:153990831 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTGTAGCTGTTAACAAAAAACTA[A/C]AACACTGTAACTACCCCAAATAAAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM114A2 PubMed Links

Gene Details

Gene
FAM114A2
Gene Name
family with sequence similarity 114 member A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317993.1 3759 UTR 3 NP_001304922.1
NM_001317994.1 3759 UTR 3 NP_001304923.1
NM_001317995.1 3759 UTR 3 NP_001304924.1
NM_018691.3 3759 UTR 3 NP_061161.2
XM_005268360.4 3759 Intron XP_005268417.1
XM_005268361.4 3759 Intron XP_005268418.1
XM_011537540.2 3759 Intron XP_011535842.1
XM_011537541.2 3759 Intron XP_011535843.1
XM_011537542.2 3759 Intron XP_011535844.1
XM_017008967.1 3759 Intron XP_016864456.1
XM_017008968.1 3759 Intron XP_016864457.1

View Full Product Details