Product Details

SNP ID
rs7290732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:46296006 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGTGCTCTCGAGTTCCATCCTTT[C/T]CAGAGCGGGTCCTCCCAGACTCCTC
Phenotype
MIM: 607477
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GTSE1 PubMed Links

Gene Details

Gene
GTSE1
Gene Name
G2 and S-phase expressed 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016426.6 126 Intron NP_057510.4
XM_005261627.3 126 UTR 5 XP_005261684.1
XM_011530211.2 126 UTR 5 XP_011528513.1
XM_011530213.2 126 UTR 5 XP_011528515.1
XM_017028815.1 126 Intron XP_016884304.1
XM_017028816.1 126 UTR 5 XP_016884305.1
Gene
GTSE1-AS1
Gene Name
GTSE1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
TTC38
Gene Name
tetratricopeptide repeat domain 38
There are no transcripts associated with this gene.

View Full Product Details