Product Details

SNP ID
rs8016694
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74662880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATGGGGAAGTCAGTGGCCCAAGC[C/T]GGCCATACTTCAGTGCCAATAACAA
Phenotype
MIM: 615380
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
AREL1 PubMed Links

Gene Details

Gene
AREL1
Gene Name
apoptosis resistant E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039479.1 3604 UTR 3 NP_001034568.1
XM_005268244.3 3604 UTR 3 XP_005268301.1
XM_006720344.2 3604 UTR 3 XP_006720407.1
XM_011537415.2 3604 UTR 3 XP_011535717.1
XM_011537416.2 3604 UTR 3 XP_011535718.1
XM_011537417.2 3604 UTR 3 XP_011535719.1

View Full Product Details