Product Details

SNP ID
rs8043721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:50248341 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGCACAAAGGAATGTTGCTGAT[A/G]GAGAGGGGCCTATGTCCCCTCCTCC
Phenotype
MIM: 600385
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADCY7 PubMed Links

Gene Details

Gene
ADCY7
Gene Name
adenylate cyclase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114.4 Intron NP_001105.1
NM_001286057.1 Intron NP_001272986.1
XM_011522835.2 Intron XP_011521137.1
XM_011522836.2 Intron XP_011521138.1
XM_011522837.2 Intron XP_011521139.1
XM_011522838.2 Intron XP_011521140.1
XM_011522839.2 Intron XP_011521141.1
XM_011522840.2 Intron XP_011521142.1
XM_011522842.2 Intron XP_011521144.1
XM_017022895.1 Intron XP_016878384.1
XM_017022896.1 Intron XP_016878385.1
XM_017022897.1 Intron XP_016878386.1
XM_017022898.1 Intron XP_016878387.1
XM_017022899.1 Intron XP_016878388.1

View Full Product Details