Product Details

SNP ID
rs7133325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:118986045 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAGTAGACTTCAGCCCAGGCCCT[C/G]TCCAAAAGGCCATAAATTCCAAACT
Phenotype
MIM: 613103
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SRRM4 PubMed Links

Gene Details

Gene
SRRM4
Gene Name
serine/arginine repetitive matrix 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194286.3 Intron NP_919262.2

View Full Product Details